Penicillin : Streptomycin solution 50X, Corning®
Supplier: Corning
|
Danger
|
This product is a mix of the antibiotics Penicillin (5,000 IU) and Streptomycin (5,000 μg/ mL) in a 50-fold working concentration. Penicillin (Penicillin G) works by inhibiting peptidoglycan synthesis, while Streptomycin inhibits protein synthesis. Penicillin-Streptomycin is effective against Gram-negative and Gram-positive bacteria. This antibiotic is recommended for use in cell culture media at 20 ml/L.
Caution: Storage -25°C to -15°C. For Research or Further Manufacturing Use Only. Not for use in Diagnostic Procedures.
CAS Number:
1406-05-9 (Penicillin) - 57-92-1 (Streptomycin) |
Specification Test Results
Formulation: | |
Penicillin G, potassium salt | 5,000,000 U/L |
Streptomycin sulfate | 5.0 g/l |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...