Neomycin sulfate, powder

Supplier: Corning

Synonyms: Neomycin, Fradiomycin sulfate, (2S,3S,4R,5R,6R)-5-Amino-2-(aminomethyl)-6-(((2R,3S,4R,5S)-5-(((1R,2R,3S,5R,6S)-3,5-diamino-2-(((2R,3R,4R,5S,6R)-3-amino-6-(aminomethyl)-4,5-dihydroxytetrahydro-2H-pyran-2-yl)oxy)-6-hydroxycyclohexyl)oxy)-4-hydroxy-2-(hydroxymethyl)tetrahydrofuran-3-yl)oxy)tetrahydro-2H-pyran-3,4-diol sulphate, Fradiomycin sulphate, Neomycin sulphate salt, Neomycin sulfate salt, (2S,3S,4R,5R,6R)-5-Amino-2-(aminomethyl)-6-(((2R,3S,4R,5S)-5-(((1R,2R,3S,5R,6S)-3,5-diamino-2-(((2R,3R,4R,5S,6R)-3-amino-6-(aminomethyl)-4,5-dihydroxytetrahydro-2H-pyran-2-yl)oxy)-6-hydroxycyclohexyl)oxy)-4-hydroxy-2-(hydroxymethyl)tetrahydrofuran-3-yl)oxy)tetrahydro-2H-pyran-3,4-diol sulfate

61-241-RG
45000-644EA 129.58 USD
45000-644
Neomycin sulfate, powder
Neomycin sulphate

Neomycin, a nucleoside antibiotic and aminoglycoside, is a protein synthesis inhibitor that causes premature chain termination by acting as an analog of the 3'-terminal end of aminoacyl-tRNA


It prevents growth of bacteria, protozoa, algae, and mammalian cells. Organisms carrying the resistance gene puromycin acetyltransferase are effectively protected from this compound. Neomycin is effective against Gram-negative and Gram-positive bacteria.


Packaging: Shipping temperature: Ambient

Formula: C₂₃H₄₆N₆O₁₃·3H₂SO₄
MW: 908.87 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150827
CAS Number: 1405-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR