Kanamycin sulfate, powder

Supplier: Corning

61-176-RG
45000-640EA 140.09 USD
45000-640
Kanamycin sulfate, powder
Kanamycin sulphate

Kanamycin is a member of the aminoglycosides a group of molecules and interacts with the 30S subunit of prokaryotic ribosomes


It induces errors in translation and thus hinders protein synthesis. The KanR-Tn5 gene product (aminoglycoside phosphotransferase) confers resistance to kanamycin. Kanamycin Sulfate is effective against Gram-negative and -positive bacteria.


Packaging: Shipping temperature: Ambient

Formula: C₁₈H₃₈N₄O₁₅S
Storage Temperature: Refrigerator
MDL Number: MFCD00070253
CAS Number: 25389-94-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR