Chloramphenicol, powder

Supplier: Corning

61-239-RI
45000-618EA 122.77 USD
45000-618
Chloramphenicol, powder
Chloramphenicol

Chloramphenicol is a common antibiotic which inhibits microbial protein synthesis by preventing peptide bond formation. It is effective against Gram-positive and Gram-negative bacteria.


Packaging: Shipping temperature: Ambient

Formula: Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂
Storage Temperature: Ambient
MDL Number: MFCD00078159
CAS Number: 56-75-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR