Corning® Hink's TNM-FH Medium, Corning
Supplier: Corning
13-100-CV
45000-478PK 426.88 USD
45000-478
Corning® Hink's TNM-FH Medium, Corning
Cell Culture Media
Hink's TNM-FH medium is a modified Grace's formulation containing lactalbumin hydrolysate (LAH) and ultrafiltered yeastolate, and was originally developed for the optimal propagation of cabbage looper, Tricoplusi in cells.