Corning® Graces's Insect Basal Medium (Vaughn Mod.), Corning
Supplier: Corning
13-200-CV
45000-476PK 386.27 USD
45000-476
Corning® Graces's Insect Basal Medium (Vaughn Mod.), Corning
Cell Culture Media
The insect cell culture medium was formulated to closely resemble insect hemolymph, a compound crucial to the function of the insect circulatory system
The basal medium, when properly supplemented, has been used to culture cells derived from a variety of insects including several species of lepidopterans as well as some dipterans. The medium is primarily used as a basal medium for the growth and maintenance of cell lines derived from lepidopterans.