Corning® insectagro® DS2 Serum-Free/Protein-Free Medium, 1X without L-glutamine, Corning
Supplier: Corning
13-402-CV
45000-474PK
229.88
USD
45000-474
Corning® insectagro® DS2 Serum-Free/Protein-Free Medium, 1X without L-glutamine, Corning
Cell Culture Media
Corning® insectagro® DS2 Serum-Free/Protein-Free Medium was developed for the growth and maintenance of Drosophila Schneider 2 (S2) cells to be used in heterologous protein expression
At the optimal temperature range 22-24ºC, DS2 cells grow as a loose monolayer and are readily adaptable to growth in suspension. Under these conditions, the cells require minimal adaptation to serum-free culture.