Corning® Serum-Free/Protein-Free Medium, 1X without L-Glutamine, Corning
Supplier: Corning
40-200-CV
45000-424PK
195.22
USD
45000-424
Corning® Serum-Free/Protein-Free Medium, 1X without L-Glutamine, Corning
Cell Culture Media
Optimized for the cultivation of Chinese Hamster Ovary (CHO) cells, along with many derivatives of this parent line, Corning® cellgro® FREE is a proprietary serum-free and protein-free growth medium that does not contain any hormones or growth factors. This medium supports superior growth and viability over long term passages in both adherent and suspension culture.