Corning® Medium 199 (Mod.), Corning
Supplier: Corning
10-060-CV
45000-570EA 119.7 USD
45000-570 45000-574 45000-378
Corning® Medium 199 (Mod.), Corning
Cell Culture Media
Medium 199 is often used in vaccine production, virology, and for the culture of various non-transformed cells
  • Low endotoxin - < 0.25EU/mL
  • Mycoplasma tested
  • Liquid media products are formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment
It was originally developed as the result of efforts to create a fully defined, animal origin-free medium. Long-term use of this medium requires serum supplementation.