Corning® Ham's F-10 Medium, Corning
Supplier: Corning
10-070-CV
45000-356PK
201.7
USD
45000-356
Corning® Ham's F-10 Medium, Corning
Cell Culture Media
Ham's F-10 is a classical media designed to support the growth of a variety of cell types, including HeLa, Chinese Hamster Ovary (CHO), primary cells from tissue explants and various types of human diploid cells.
- Low endotoxin - < 0.25EU/mL
- Mycoplasma tested
- Formulated with cell culture grade WFI water
- Manufactured in a cGMP environment