Corning® F-12K Nutrient Mixture (Kaighn's Mod.) with L-glutamine, Corning
Supplier: Corning
10-025-CV
45000-354PK
283.22
USD
45000-354
Corning® F-12K Nutrient Mixture (Kaighn's Mod.) with L-glutamine, Corning
Cell Culture Media
A modification of Ham's F-12, F-12K contains higher concentrations of amino acids, sodium pyruvate and other components than the original
- Low endotoxin: >0.25EU/mL
- Mycoplasma tested
- Formulated with cell culture grade WFI water
- Manufactured in a cGMP environment
It is formulated to support the growth of cells including differentiated rat and chicken cell types and primary human hepatocytes.