Nunc® MicroWell™ 96-Well Plates, Thermo Scientific
Supplier: Thermo Fisher Scientific
237105 237107 236108 236105
43300-448CS 380.96 USD
43300-448 43300-450 43300-438 43300-434
Nunc® MicroWell™ 96-Well Plates, Thermo Scientific
Microplates Immunology Microplates and Strips
Black and white 96-well plates are specifically designed for fluorescence and luminescence assays
Nunclon™Δ treated surface ensures optimal cell adhesion and growth. Non-treated surface is ideal for use in liquid phase immunoassays, serological tests and other homogeneous assays. MaxiSorp™ surface has a high affinity for proteins, especially antibodies. PolySorp™ surface is less polar than MaxiSorp and has an affinity towards molecules with mainly hydrophobic domains. MaxiSorp™ and PolySorp™ surfaces are used in many solid phase immunoassays.
Made of polystyrene, they feature a flat-bottom well design and have a total well volume of 400µL with a recommended working volume of 200 to 350µL. Select resins and manufacturing methods ensure maximum signal, low background, and minimal signal crosstalk. Alphanumeric coordinates offer easy orientation and sample identification. Meet the proposed SBS industry guidelines.
Ordering information: Plates are packaged 10 per sleeve with the exception of 43300-434, which is packed 1 per sleeve.