8211 N95 Particulate Respirator, 3M™
Supplier: 3M
7000029720
43100-118CS 474.82 USD
43100-118
8211 N95 Particulate Respirator, 3M™
Respirators Disposable Respirators Particulate Respirators
Designed to help provide comfortable reliable worker protection, this respirator can be used for grinding, sanding, sweeping, bagging, and other dusty operations.
  • Provide comfortable reliable worker protection in grinding, sanding, and other dusty operations
  • 3M Cool Flow® Exhalation Valve, adjustable “M” nose clip, conformable face seal, and durable braided headband
  • Ideal for work settings that involve heat, humidity, or long periods of wear
  • Provides minimum 95% filtration efficiency against certain solid and liquid particles that do not contain oil
Features the 3M Cool Flow* Exhalation Valve, adjustable “M” nose clip, comfortable faceseal, and durable braided headband. Ideal for work settings that involve heat, humidity, or long periods of wear. Provides minimum 95% filtration efficiency against certain solid and liquid particles that do not contain oil.
Caution: All 3M™ products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.