1860 Disposable N95 Health Care Particulate Respirator and Surgical Masks 3M™
Supplier: Healthcare Products
MMM 1860
43100-054CS 390.38 USD
43100-054 43100-064
1860 Disposable N95 Health Care Particulate Respirator and Surgical Masks 3M™
Respirators Disposable Respirators Particulate Respirators
This respirator has a soft inner shell for extended comfort on the face.
  • Fluid-resistant to reduce the potential exposure of the wearer to blood and other body fluids
  • Provides minimum filtration efficiency of 95% against certain solid and liquid particles that do not contain oil
Meets CDC guidelines for TB exposure control and helps reduce exposure to airborne particles.
Caution: All 3M™ products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.