Fixation Buffer, Biolegend
Supplier: Biolegend
420801
420801-BLEA 86.6 USD
420801-BL
Fixation Buffer, Biolegend
Buffers Microscopy Buffers
Fixation Buffer is useful for intracellular staining procedures like preparation of cells for staining intracellular cytokines or other proteins and to fix cells prior to permeabilization using permeabilization wash buffer.