Hematoxylin 1, Signature Series™

Supplier: Epredia

7221
41810-462CS 485.16 USD
41810-462
Hematoxylin 1, Signature Series™
Hematoxylin

Epredia™ Hematoxylin 1 is a progressive stain developed to be used as a nuclear stain in the Hematoxylin and Eosin-Y staining procedure and in cytology. The stain requires no filtration prior to use and will not form crystals upon standing or exposure to cold weather.


  • Differentiation is not necessary, however, Clarifier 1 should be used to eliminate any background staining caused by use of excessive water bath adhesive
  • Rapid stain requires no filtration prior to use
  • Will not form crystals upon standing or exposure to cold weather

Formula: C₁₆H₁₄O₆
MDL Number: MFCD00078111
CAS Number: 517-28-2
IVDLogo

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR