Hematoxylin 2, Signature Series™

Supplier: Epredia

7231
41810-446CS 504.9 USD
41810-446
Hematoxylin 2, Signature Series™
Hematoxylin

Richard-Allan Scientific Signature Series Hematoxylin 2 produces staining intensity similar to Harris Hematoxylin.


  • Unlike many comparable products, Hematoxylin 2 does not contain mercuric oxide and will not form a metallic sheen over its surface each day
  • Differentiation is not necessary, however, Clarifier 2 should be used to eliminate any background staining caused by the use of excessive water bath adhesive
  • Recommended for frozen sections
  • Requires no filtration prior to use
  • Will not form crystals upon standing or exposure to cold weather

Formula: C₁₆H₁₄O₆
MDL Number: MFCD00078111
CAS Number: 517-28-2
IVDLogo

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR