VWR® Disposable Aspirating Pipettes, Polystyrene, Sterile
Supplier: VWR International
414004-264 414004-265 414004-266 414004-267 414004-268
414004-264CS 69.87 USD
414004-264 414004-265 414004-266 414004-267 414004-268
VWR® Disposable Aspirating Pipettes, Polystyrene, Sterile
Pipettes Serological Pipettes
VWR® Disposable Aspirating Pipettes are manufactured from polystyrene and supplied sterile.
  • Non-plugged, nongraduated, single-use pipettes
  • Thermoformed paper/plastic packaging protects against contamination
These non-plugged, nongraduated, single-use pipettes can be used for a wide variety of vacuum-aspirating procedures, and are a safer alternative to glass pasteur pipettes. Pipettes are supplied in a thermoformed paper/plastic packaging wrap to protect against contamination. Nonpyrogenic.
Packaging: Pipettes are supplied individually wrapped inMore Product Information