VWR® Large Square Wide Mouth Bottle, PP
Supplier: VWR International
414004-189
414004-189CS 274.98 USD
414004-189
VWR® Large Square Wide Mouth Bottle, PP
Bottles Plastic Bottles
Space-saving 3.8 L (1 gal.) bottle is ideal for storing and sterilizing contaminated slides and other small lab items.
  • Autoclavable
  • Integrated handle for improved grip and convenience
  • Not leakproof
  • Polypropylene construction with wide mouth
Features integrated handle for better grip, greater safety, and pouring convenience.
Note: Before autoclaving, set cap on top of bottle without engaging threads.
Ordering information: Size 100-415 white polypropylene screw cap included, assembled.
Caution: Bottle is autoclavable, but not leakproof.