VWR® Mechanical Pump Oil No. 21
Supplier: VWR International
40300-028
40300-028CS 675.7 USD
40300-028
VWR® Mechanical Pump Oil No. 21
Lubricants
Designed for use in applications where reactive or corrosive gases are present.
  • Resistant to oxidation and decomposition
  • Good vapor pressure capabilities
Additives resist oxidation and decomposition. The oil uses molecular distillation to ensure good vapor pressure capabilities. Vapor Pressure at 25 °C is 3e-5torr. Boiling Point at 0.01torr is 112 °C.