VWR® CERTICLEAN® Cleanroom Latex Gloves, Class 10, Hand-Specific
Supplier: VWR International
40101-044 40101-046 40101-048 40101-052 40101-054 40101-056 40101-058
40101-044CS 947.06 USD
40101-044 40101-046 40101-048 40101-052 40101-054 40101-056 40101-058
VWR® CERTICLEAN® Cleanroom Latex Gloves, Class 10, Hand-Specific
Gloves
Hand-specific gloves with beaded cuffs for strength and reduction of cuff roll down.
  • Powder-free, natural rubber latex
  • Tapered, beaded cuff
  • Certificates of analysis available
  • Quality data available
The length and tapered cuff provide more coverage over the garment sleeve.
Uncompromising manufacturing processes consistently yield exceptionally low particle counts. Specialized manufacturing processes and silicone-free material minimize ionic contaminants and nonvolatile residue (NVR) transfer.
Film thickness designed for comfort and tactile sensitivity. Textured for a reliable grip.
Certifications: Certificates of Analysis Available. Quality Data Available for Product Specifications, Test Methods, Historical Trend Analysis and Contamination Specifications.
Ordering information: Packed in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom. Packaged in cleanroom-compatible polyethylene, 50 pair per double poly bag, 4 bags per case.
Caution: This product contains natural rubber latex which may cause allergic reactions. Safe use of this glove by or on latex-sensitive individuals has not been established. This product should not be worn by or exposed to individuals allergic to natural rubber latex.