Anti-IgG2b, k Isotype Ctrl Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: MPC-11]

Supplier: Biolegend
400334
400334-BLEA 217.3 USD
400334-BL
Anti-IgG2b, k Isotype Ctrl Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: MPC-11]
Antibodies
Alexa Fluor® 700 Mouse IgG2b, κ Isotype Ctrl [MPC-11]; Isotype: Mouse IgG2b, κ; Reactivity: Mouse; Apps: FC, ICFC; Size: 100 μg

This reagent is developed for immunofluorescent staining for flow cytometric analysis as negative control. Use at concentrations comparable to those of the specific antibody of interest. It is highly recommended that the reagent be titrated for optimal performance for each application.

Type: Control
Antigen: IgG2b, k Isotype Ctrl
Clonality: Monoclonal
Clone: MPC-11
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR