Precurved Anti-Vibration/Impact Gloves, Half-Finger, OK-1® OccuNomix
Supplier: Occunomix International, LLC
OK980-L XXL
39870-218CS 485.11 USD
39870-218 39870-226 39870-228
Precurved Anti-Vibration/Impact Gloves, Half-Finger, OK-1® OccuNomix
Gloves
These fully washable leather antivibration gloves feature a precurved shape and half finger design for dexterity.
  • Gloves provide superior protection from sources of vibration and impact
  • Can be washed and dried as needed to prolong their llfe and appearance
They help you maintain a consistent grip because they are curved to match the natural shape of the hand.
Ordering information: Must specify left or right hand when ordering.