Universal® Back Support Belt, OK-1® Safety, OccuNomix
Supplier: Occunomix International, LLC
OK-UNIV
39870-080CS 992.65 USD
39870-080
Universal® Back Support Belt, OK-1® Safety, OccuNomix
Ergonomic Supports Back Supports
Designed for moderate lifting requirements
The Universal back support belt with suspenders features a detachable swivel clip system and will fit waist sizes 66–122cm (26–48"). It features a double-overlapping closure system for great support and an open mesh body to help reduce heat build-up. Two-way stretch elastic is doubled-up for greater support. Interior rubber tracks and plastic stays help the belt stay in place.
Ordering information: For belts without suspenders, contact your Avantor sales representative.