ExStik® Waterproof pH Meter, Extech
Supplier: NEW GENIE GROUP INC SE
PH100
39000-094EA 229.9 USD
39000-094
ExStik® Waterproof pH Meter, Extech
Electrochemistry Meters pH and Multi-Parameter Meters
Meter with flat surface electrode for fast and easy on-the-spot pH measurements
The PH100 features a built-in Smart chip offers advanced functions and features to a stick style pH meter not found anywhere else. It also has a RENEW feature that tells you when it’s time to replace your pH electrode and a CAL feature that tells you when it’s time to recalibrate your meter. Other features include a Flat Surface pH electrode that is completely waterproof and rugged enough to withstand harsh environments.
Certifications: CE marked. IP57.
Ordering information: One year warranty (meter); 6 months conditional (electrode). Complete with gel filled flat surface pH electrode, sensor cap, sample cup with cap, four 3V CR2032 buttome batteries, and 48" neckstrap.