Erem® Magic™ Diagonal Cutter, Semi-Flush Cut, Cooper Tools
Supplier: Cooper Tools
595T
37699-350EA 622.32 USD
37699-350
Erem® Magic™ Diagonal Cutter, Semi-Flush Cut, Cooper Tools
Cutters
This tapered head side cutter is designed with tungsten carbide blades. The jaws of the cutter have straight edges and taper to a point.
This tapered head shape allows access to difficult to reach areas but reduces the cutting capacity in comparison to the same size oval head cutter.