Weller* Standard Series Modular Heater for Thread-On Tips
Supplier: APEX TOOL GROUP
50W heater. Temperature: 537[degree]C. Use with Weller* Standard Series Modular Iron Handles (37698-964, -962, -960).