Weller* Controlled-Output Iron
Supplier: APEX TOOL GROUP
Portable. 60W. Closed loop method controls maximum tip temperature. Stainless steel modular plug-in heater for greater efficiency and versatility. With 371[degree] screwdriver tip 37691-248, nichrome wound heating element, 1.8m 3-wire cord, stand. 120V. UL/C-UL.