Weller® Contact Pyrometer, Apex
Supplier: Cooper Tools
CP01
37697-504EA 108.73 USD
37697-504
Weller® Contact Pyrometer, Apex
Soldering Supplies Soldering Stations
Contact pyrometer with large 20-gauge wire for use with WA2000 soldering iron analyzer.
  • Quickly measures the temperature stability of a soldering iron tip