Kahnetics® Tapered Needles, Cooper Tools
Supplier: Cooper Tools
KDS14TNP KDS16TNP KDS18TNP KDS22TNP
37696-952EA 100.23 USD
37696-952 37696-998 37695-028 37695-148
Kahnetics® Tapered Needles, Cooper Tools
All-polyethylene dispensing tips
Tapered design reduces dripping. Polyethylene material won’t scratch delicate surfaces. Overall length: 38.1mm (11/2").
Ordering information: Tips are packaged 50 per pack.