Kahnetics* Tapered Needle
Supplier: APEX TOOL GROUP
All-polyethylene dispensing tip. Tapered design reduces dripping. Polyethylene material will not scratch delicate surfaces. Overall length: 38.1mm. Color: salmon. Gauge: 14. I.D.: 1.60mm.