Heat Gun, Light Weight, Cooper Tools
Supplier: Cooper Tools
6966C
37696-762EA 1020.39 USD
37696-762
Heat Gun, Light Weight, Cooper Tools
Heaters Hot Air Dryers
This ESD safe heat gun is perfect for repetitive or continuous use in production plants, labs, and electronics service facilities
  • Heavy duty heater and fan components for long service life
  • Light weight for reduced operator fatigue
  • Perfect for shrink tubing and vinyl/T.F.E. epoxy curing, drying, and cooling
  • Three-position switch: HOT – OFF – COOL
Certifications: UL and cUL Listed.