Weller® UNGAR® Flux Solvent Dispenser, Cooper Tools
Supplier: Cooper Tools
FD2
37695-400EA 25.31 USD
37695-400
Weller® UNGAR® Flux Solvent Dispenser, Cooper Tools
Bottles Dropping and Dispensing Bottles
Avoids spills and waste and places flux precisely where needed.
Simply squeeze bottle to dispense one drop of liquid resin, flux, or oil solvent through stainless steel needle.