Weller* Soldering Iron Tip
Supplier: APEX TOOL GROUP
Screwdriver tip for 371[degree]C soldering. For use with irons 37698-666 and -668. Copper core electroplated with iron. Nickel and chromium behind working surface protect shank from corrosion and solder creep. Maximum heat transfer. Reach: 34.92mm. Size: 3.17mm.