Xcelite* Magnetic Nutdriver
Supplier: APEX TOOL GROUP
Alnico permanent magnet in socket holds fasteners firmly for easy, one-handed driving; also retrieving and removing. Socket treated and hardened for use with self-tapping screws. Contoured red handle. Diameter: 6.4mm. Blade length: 15.2cm. Overall length: 25.7cm.