Weller* Soldering Iron Tip
Supplier: APEX TOOL GROUP
Long screwdriver tip for 427[degree]C soldering. Copper core electroplated with iron. For use with iron 37698-466 or station 37697-832. Nickel and chromium behind working surface protect shank from corrosion and solder creep. Reach: 25.4mm. Size: 3.17mm.