Vibrax™ VXR Orbital Shaker, 230 V
Supplier: IKA Works
2819000
37002-064EA 2223.36 USD
37002-064
Vibrax™ VXR Orbital Shaker, 230 V
Shakers and Mixers Orbital Shakers
Suitable for applications requiring continuous operation.
  • 230 V model
Universal platform features rollers for rapid and secure clamping of vessels at least 8 cm high. Universal platform dimensions: 26.5×13.6×6 cm (10⁷/₁₆×5³/₈×2³/₈"). The one-hand platform is for shaking single, non-fixed vessels sized 1 to 250 ml. One-hand platform dimensions: 13×13.5×4 cm (5¹/₈×5⁵/₁₆×1¹/₂"). The centrifuge tube platform is for intensive mixing of up to sixty-four 1.5mL tubes. Centrifuge tube platform dimensions: 21×21×6.5 cm (8¹/₄×8¹/₄×2¹/₂"). The dish platform is for careful mixing of culture bottles, petri dishes, and other dishes. Dish platform surface dimensions: 41×21 cm (16¹/₈×8¹/₄"). Test tube rack holds up to 36 test tubes or centrifuge tubes with a diameter of 16mm. Test tube rack dimensions: 14×14.5×11.5 cm (5¹/₂×5³/₄×4¹/₂").
Optoelectronically controlled shaker maintains slow speeds and features a smooth start to prevent splashing. Shaker also offers a small footprint and a wide speed range for a variety of applications.
Caution: 230 V model. Corresponding model with voltage suitable for use in US (37002-066) is also available, please see separate product page.