InterGuard™ Sleeve, InterWorld™
Supplier: Interworld
10135-102-02
37002-062CS 343.34 USD
37002-062
InterGuard™ Sleeve, InterWorld™
Disposable Garments
Sleeve is designed to provide barrier protection in dry applications
It offers a secure fit with elastic at both ends. Constructed of lightweight, breathable, spunbonded polypropylene.
Ordering information: Packaged 100 sleeves per bag, 3 bags per case.