7701™ Prism® Primer, Loctite, Henkel
Supplier: Loctite
135449
37001-751CS 524.13 USD
37001-751
7701™ Prism® Primer, Loctite, Henkel
Adhesives
This primer is used to make polyolefin and other low energy surfaces suitable for bonding with cyanoacrylate adhesives.
  • Colorless
  • Solvent-based liquid primer
It is only recommended for difficult-to-bond substrates which include PE, PPL, PTFE and thermoplastic rubber materials. It is not recommended where high peel strength is required. It is suitable for use in the assembly of disposable medical devices.
Certifications: ISO 10993 approved.