Nunc™ Polyolefin Acrylate Sealing Tape, Thermo Scientific
Supplier: Thermo Fisher Scientific
232702
37000-548CS 134.96 USD
37000-548
Nunc™ Polyolefin Acrylate Sealing Tape, Thermo Scientific
Seals Microplate Seals
The polyolefin acrylate sealing tapes minimize evaporation and protect samples from contamination and spilling.
  • Use for PCR, fluorescence assays, and microscopy applications
  • Pressure sensitive adhesive
  • Non-pierceable
  • Nonsterile
Ideal for all assay applications.
Certifications: Certified RNase- and DNase-free.
Ordering information: Compatiable with 96- and 384-well plates.