Steam Generator, Koehler
Supplier: Koehler
K33850
37000-244EA 14784.35 USD
37000-244
Steam Generator, Koehler
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
Electrically heated boiler provides instantaneous and reserve steam capacity for steam-jet evaporation tests. Requires only a water feed source and electrical hook-up.
  • For steam-jet method testing of aircraft turbine fuels
  • Meets output requirements of three-unit and six-unit evaporation baths
  • Electrically heated for clean, efficient operation and ease of installation
Ruggedly constructed, with long life industrial grade heating element. Includes a full range of safety features such as automatic water level control and low water cut-off, steam safety valve, high-limit pressure cut-out with manual reset and steam pressure gauge.
Produces 54.1 lbs. steam/hr at 212 °F.
Certifications: Meets applicable ASME, NEC standards.