VWR® Traceable® Infrared Thermometer with Pocket Clip
Supplier: VWR International
36934-174
36934-174EA 94.4 USD
36934-174
VWR® Traceable® Infrared Thermometer with Pocket Clip
Thermometers Digital Thermometers
Noninvasive, no-touch measurements are ideal for food preparation, life sciences, pharmaceuticals, petroleum products, cleanrooms, electronics, and field use.
  • Traceable®
  • Simple point-and-shoot operation
  • Pocket-sized
Unit takes readings in less than a second
Field of view is 1:1; thermometer can read a spot the size of 5 cm (2") from 5 cm (2") away. The fixed emissivity of 0.95 covers most applications. Features a handy pocket clip, key-chain, and lanyard. Display indicates low battery.
Unit takes both Celsius and Fahrenheit readings of solids, semi-solids, and liquids. Hold freezes the display at the current reading.
Certifications: A serial-numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with button cell battery.