VWR® Traceable® Hygrometer/Thermometer
Supplier: VWR International
36934-164 10754-738 10754-854
36934-164EA 140.62 USD
36934-164 10754-738 10754-854
VWR® Traceable® Hygrometer/Thermometer
Hygrometers Digital Thermo-Hygrometers
Displays current, minimum, and maximum values for dew point, wet-bulb, humidity, and temperature.
  • High/low alarm options
  • Bright LCD digits
  • Chemical-resistant, high-impact ABS plastic case
  • Accurate temperature and water content readings
High/low alarms may be set in 0.1 increments for all four readings. Alarm sounds for one minute and a red LED flashes until reading returns to non-alarm conditions. LCD digits are 2.5 cm (1”) high. Case is constructed from high-impact, chemical-resistant ABS plastic.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology. Monitor also meets CLIA specification 493.1207 for monitoring temerature and humidity.
Ordering information: Supplied with two AA batteries.