VWR® Traceable® Big-Digit See-Thru™ Thermometers
Supplier: VWR International
36934-156 36934-158
36934-156EA 70.57 USD
36934-156 36934-158
VWR® Traceable® Big-Digit See-Thru™ Thermometers
Thermometers Digital Thermometers
Use outdoors and in storerooms, fume hoods, labs, cleanrooms, refrigerators, and plant areas.
  • Window or wall mount
  • Automatic minimum/maximum reset
  • Transparent display
  • Temperatures automatically cleared and updated daily
Thermometer may be attached to the outside of a window to view outdoor temperatures. Features a transparent display with 3.2 cm (11/4") high digits to show current and minimum/maximum temperatures, which are automatically cleared and updated daily.
Certifications: A serial-numbered certificate is provided from an ISO 17025More Product Information