VWR® Dust'it™ Blower
Supplier: VWR International
36934-142
36934-142EA 22.76 USD
36934-142
VWR® Dust'it™ Blower
Sprays
Providing high-pressure, pinpoint jets of air, this powerful bulb duster forcibly blows away dust, lint, and dirt.
  • Forcibly blows away dust, lint, and dirt
  • Ideal for delicate, hard-to-clean areas
  • Nonconductive plastic sleeve tip prevents scratches to surfaces
  • One-way inlet valve at the top keeps air from being drawn in
Ideal for delicate, difficult-to-clean, and inaccessible equipment. The nonconductive plastic sleeve tip prevents scratches for cleaning optical lenses, slides, circuit boards, and cuvets. The one-way inlet valve at the top stops air from being drawn through the tip. Varying the “squeeze” provides for controlled delivery.