VWR® Traceable® Econo Refrigerator Thermometer
Supplier: VWR International
36934-132
36934-132EA 49.62 USD
36934-132
VWR® Traceable® Econo Refrigerator Thermometer
Thermometers Digital Thermometers
Economical, lightweight thermometers are ideal for monitoring temperatures in storerooms, labs, offices, and hoods.
  • Transparent LCD display
  • Economical
  • Lightweight
  • Switchable °C/°F
The see-through LCD updates every 30 seconds and features large 1.3 cm (1/2") digits
A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Included attachment devices allow for flexible positioning.
Ordering information: Supplied with a long-life 1.5V AG10 silver oxide battery, Always-hold™ suction cup, and Velcro® fastener.