VWR® Large Cylindrical Scoops, PE
Supplier: VWR International
36760-6000
36494-110EA 103.29 USD
36494-110 70680-280
VWR® Large Cylindrical Scoops, PE
Scoops
Large, heavy-duty, polyethylene plastic scoops are strong, durable, and excellent for scooping dry powders, granules, or crystals.
  • Closed back, non-spill shape
  • Ergonomically angled handle