Nalgene® Cryoware Labels, Thermo Scientific
Supplier: Thermo Fisher Scientific
5040-0002
36471-703CS 1280.56 USD
36471-703
Nalgene® Cryoware Labels, Thermo Scientific
Labels
Specifically designed for use at ultra-low temperatures.
  • Durable
  • Will not peel or shrink, even upon removal from mechanical or liquid-nitrogen freezers
Cloth labels adhere to plastic and cardboard cryogenic storage boxes.
Accessories information: Marker set contains four colors: red, blue, black, and green.
Ordering information: Labels and marker set are sold separately.