Nalgene® PolyPaper® Right-To-Know Custom Labeling System, Thermo Scientific
Supplier: Thermo Fisher Scientific
6316-1000
36471-606CS 1244.81 USD
36471-606
Nalgene® PolyPaper® Right-To-Know Custom Labeling System, Thermo Scientific
Labels
Pressure-sensitive labels incorporate Department of Transportation and National Fire Protection Association codes, symbols, and pictorials of recommended safety equipment in an easily recognizable format.
  • Made of tear-resistant NALGENE® PolyPaper
  • Waterproof and chemical resistant
Custom Labeling System includes pressure sensitive stickers depicting NFPA rating numbers, hazard codes, and safety pictorials to be placed on the blank PolyPaper Label.
A Mylar® overlay is provided to protect the label from chemicals. A special adhesive backing adheres to any plastic, glass or metal transfer container.
Certifications: Assures compliance with the OSHA Hazard Communication Regulation CFR 29 1910.1200 (f)(4).