Shake ‘N’ Bake Hybridization Oven, Boekel Scientific
Supplier: Boekel
136400
35821-077EA 5667.74 USD
35821-077
Shake ‘N’ Bake Hybridization Oven, Boekel Scientific
Ovens Hybridisation Ovens
This hybridization incubator is ideal for hybridizing and prehybridizing membranes used in Southern and Northern blotting techniques
Three 22x20x1.3cm (821/32x77/8x1/2") shelves, spaced 6.35cm (21/2") apart, accommodate dishes or heat-sealed bags. Shelves are removable to increase space for taller vessels. Equipped with a gasket-sealed, metal and acrylic see-through door.
The control panel features temperature selection buttons; an easy-to-read LCD temperature display; an independent, calibrated, safety thermostat; and a speed adjustment knob.
The gentle rocking motion provides washes that are less messy than a shaking water bath and more convenient than a shaker inside a conventional incubator. The small footprint conserves valuable bench space.
Ordering information: Supplied with manufacturer’s two-year warranty.